3-(Benzyloxy)-5-bromo-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48951-500MG 48951-5G 48951-25G
77484-148EA 202.91 USD
77484-148 77494-632 77504-218
3-(Benzyloxy)-5-bromo-2-fluorobenzaldehyde ≥97%
3-(Benzyloxy)-5-bromo-2-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD33403924
CAS Number: 2624417-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR