3,6-Dichloro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

48865-500MG 48865-5G 48865-25G
77484-140EA 160.68 USD
77484-140 77494-624 77504-210
3,6-Dichloro-2-(methylthio)phenol ≥97%
3,6-Dichloro-2-(methylthio)phenol

Precaución: For research use only

Formula: C₇H₆Cl₂OS
MW: 209.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786656
CAS Number: 29237-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR