4-(Benzyloxy)-3-chloro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48812-500MG 48812-5G 48812-25G
77484-136EA 243.08 USD
77484-136 77494-620 77504-206
4-(Benzyloxy)-3-chloro-2-methylbenzoic acid ≥97%
4-(Benzyloxy)-3-chloro-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00598349
CAS Number: 2586125-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR