4-[4-(Hydroxymethyl)phenyl]pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-Pyridin-4-ylphenyl)methanol, (4-(Pyridin-4-yl)phenyl)methanol

48792-5G 48792-25G
77494-616EA 244.11 USD
77494-616 77504-202
4-[4-(Hydroxymethyl)phenyl]pyridine ≥95%
4-[4-(Hydroxymethyl)phenyl]pyridine

Precaución: For research use only

Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114576
CAS Number: 217192-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR