5-Bromo-1-ethyl-2-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48715-500MG 48715-5G 48715-25G
77484-130EA 166.86 USD
77484-130 77494-610 77504-196
5-Bromo-1-ethyl-2-iodo-3-methylbenzene ≥97%
5-Bromo-1-ethyl-2-iodo-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀BrI
MW: 324.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547778
CAS Number: 1208075-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR