3,6-Dichloro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

48689-500MG 48689-5G 48689-25G
77484-126EA 136.99 USD
77484-126 77494-606 77504-192
3,6-Dichloro-2-iodophenol ≥97%
3,6-Dichloro-2-iodophenol

Precaución: For research use only

Formula: C₆H₃Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD28786692
CAS Number: 63086-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR