2,6-Dichloro-3-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48655-500MG 48655-5G 48655-25G
77484-122EA 231.75 USD
77484-122 77494-602 77504-188
2,6-Dichloro-3-ethylbenzaldehyde ≥97%
2,6-Dichloro-3-ethylbenzaldehyde

Precaución: For research use only

Formula: C₉H₈Cl₂O
MW: 203.065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23163358
CAS Number: 1268865-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR