2-(5-Bromo-2-fluoro-3-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

48535-500MG 48535-5G 48535-25G
77484-118EA 181.28 USD
77484-118 77494-596 77504-182
2-(5-Bromo-2-fluoro-3-methylphenyl)propan-2-ol ≥95%
2-(5-Bromo-2-fluoro-3-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD32902744
CAS Number: 1522984-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR