2-Chloro-5-methyl-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

48418-500MG 48418-5G
77484-114EA 595.34 USD
77484-114 77494-590
2-Chloro-5-methyl-3-(methylthio)pyridine ≥97%
2-Chloro-5-methyl-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₇H₈ClNS
MW: 173.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28060981
CAS Number: 1823926-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR