2-Fluoro-3-hydroxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

48402-500MG 48402-5G
77484-110EA 511.91 USD
77484-110 77494-586
2-Fluoro-3-hydroxyisonicotinaldehyde ≥97%
2-Fluoro-3-hydroxyisonicotinaldehyde

Precaución: For research use only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611049
CAS Number: 1227594-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR