2-Bromo-6-chloro-3-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48333-500MG 48333-5G 48333-25G
77484-102EA 296.64 USD
77484-102 77494-578 77504-172
2-Bromo-6-chloro-3-isopropoxybenzaldehyde ≥95%
2-Bromo-6-chloro-3-isopropoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756430
CAS Number: 2376271-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR