1-Bromo-2-chloro-4-ethoxy-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

48069-500MG 48069-5G 48069-25G
77484-094EA 141.11 USD
77484-094 77494-570 77504-166
1-Bromo-2-chloro-4-ethoxy-5-fluorobenzene ≥97%
1-Bromo-2-chloro-4-ethoxy-5-fluorobenzene

Precaución: For research use only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34756411
CAS Number: 2386569-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR