(2-Fluoro-3-methoxypyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

48047-500MG 48047-5G
77484-090EA 509.85 USD
77484-090 77494-566
(2-Fluoro-3-methoxypyridin-4-yl)methanol ≥97%
(2-Fluoro-3-methoxypyridin-4-yl)methanol

Precaución: For research use only

Formula: C₇H₈FNO₂
MW: 157.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611249
CAS Number: 1227564-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR