(2-Bromophenyl)(o-tolyl)methanol ≥97%

Supplier: AOBChem USA

Warning

47760-500MG 47760-5G 47760-25G
77484-082EA 243.08 USD
77484-082 77494-558 77504-152
(2-Bromophenyl)(o-tolyl)methanol ≥97%
(2-Bromophenyl)(o-tolyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12964429
CAS Number: 1029874-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR