tert-Butyl (4-bromo-2-hydroxybenzyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl 4-bromo-2-hydroxybenzylcarbamate

47740-500MG 47740-5G 47740-25G
77484-078EA 232.78 USD
77484-078 77494-554 77504-148
tert-Butyl (4-bromo-2-hydroxybenzyl)carbamate ≥95%
tert-Butyl (4-bromo-2-hydroxybenzyl)carbamate

Precaución: For research use only

Formula: C₁₂H₁₆BrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32862139
CAS Number: 1402664-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR