3-(Benzyloxy)-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47722-500MG 47722-5G 47722-25G
77484-074EA 222.48 USD
77484-074 77494-550 77504-144
3-(Benzyloxy)-2-methylbenzaldehyde ≥97%
3-(Benzyloxy)-2-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22041841
CAS Number: 918524-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR