3-(tert-Butyldimethylsilyl)-1-phenylprop-2-yn-1-ol ≥95%

Supplier: AOBChem USA

Warning

476485-500MG 476485-5G 476485-25G
77484-070EA 99.91 USD
77484-070 77494-546 77504-140
3-(tert-Butyldimethylsilyl)-1-phenylprop-2-yn-1-ol ≥95%
3-(tert-Butyldimethylsilyl)-1-phenylprop-2-yn-1-ol

Precaución: For research use only

Formula: C₁₅H₂₂OSi
MW: 246.4200 g/mol
Storage Temperature: Ambient
CAS Number: 887588-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR