N-(3-Iodo-5-methylpyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

47415-500MG 47415-5G
77484-054EA 410.97 USD
77484-054 77494-530
N-(3-Iodo-5-methylpyridin-2-yl)pivalamide ≥97%
N-(3-Iodo-5-methylpyridin-2-yl)pivalamide

Precaución: For research use only

Formula: C₁₁H₁₅IN₂O
Storage Temperature: Ambient
MDL Number: MFCD16628222
CAS Number: 1228666-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR