(2,5-Difluorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

473368-500MG 473368-5G 473368-25G
77484-050EA 93.73 USD
77484-050 77494-524 77504-128
(2,5-Difluorophenyl)(methyl)sulfane ≥95%
(2,5-Difluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₆F₂S
Storage Temperature: Ambient
CAS Number: 54378-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR