(6-Bromo-2-fluoro-3-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

47297-500MG 47297-5G 47297-25G
77484-046EA 296.64 USD
77484-046 77494-520 77504-126
(6-Bromo-2-fluoro-3-(methylthio)phenyl)methanol ≥97%
(6-Bromo-2-fluoro-3-(methylthio)phenyl)methanol

Precaución: For research use only

Formula: C₈H₈BrFOS
MW: 251.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756362
CAS Number: 2807467-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR