1-Bromo-3-chloro-2-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

47262-500MG 47262-5G 47262-25G
77484-042EA 263.68 USD
77484-042 77494-514 77504-120
1-Bromo-3-chloro-2-fluoro-5-iodobenzene ≥97%
1-Bromo-3-chloro-2-fluoro-5-iodobenzene

Precaución: For research use only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30721705
CAS Number: 1416241-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR