(E)-2-Benzylidenecyclobutanone ≥95%

Supplier: AOBChem USA

Warning

469259-500MG 469259-5G
77484-038EA 382.13 USD
77484-038 77494-510
(E)-2-Benzylidenecyclobutanone ≥95%
(E)-2-Benzylidenecyclobutanone

Precaución: For research use only

Formula: C₁₁H₁₀O
Storage Temperature: Ambient
CAS Number: 208652-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR