1-(6-Methylbenzo[d][1,3]dioxol-5-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

467272-500MG 467272-5G
77484-032EA 408.91 USD
77484-032 77494-502
1-(6-Methylbenzo[d][1,3]dioxol-5-yl)propan-1-one ≥95%
1-(6-Methylbenzo[d][1,3]dioxol-5-yl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₁₂O₃
MW: 192.211 g/mol
Storage Temperature: Ambient
CAS Number: 1516523-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR