1,2-Dichloro-4-fluoro-3-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46725-500MG 46725-5G 46725-25G
77484-030EA 127.72 USD
77484-030 77494-500 77504-110
1,2-Dichloro-4-fluoro-3-(methylsulfonyl)benzene ≥95%
1,2-Dichloro-4-fluoro-3-(methylsulfonyl)benzene

Precaución: For research use only

Formula: C₇H₅Cl₂FO₂S
MW: 243.083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403841
CAS Number: 2635937-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR