2-(5-Methoxybenzo[d][1,3]dioxol-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

467195-500MG 467195-5G
77484-026EA 491.31 USD
77484-026 77494-496
2-(5-Methoxybenzo[d][1,3]dioxol-4-yl)propan-2-ol ≥95%
2-(5-Methoxybenzo[d][1,3]dioxol-4-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
CAS Number: 1148136-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR