4-Bromofuran-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

46638-500MG 46638-5G
77484-022EA 325.48 USD
77484-022 77494-492
4-Bromofuran-3-carboxylic acid ≥95%
4-Bromofuran-3-carboxylic acid

Precaución: For research use only

Formula: C₅H₃BrO₃
MW: 190.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32738839
CAS Number: 116779-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR