4-Nitro-1,3-benzothiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Nitrobenzo[d]thiazol-2-amine

465097-500MG 465097-5G 465097-25G
77484-018EA 91.67 USD
77484-018 77494-488 77504-106
4-Nitro-1,3-benzothiazol-2-amine ≥95%
4-Nitro-1,3-benzothiazol-2-amine

Precaución: For research use only

Formula: C₇H₅N₃O₂S
MW: 195.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00819610
CAS Number: 6973-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR