1-(2-Fluoro-5-methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

464732-500MG 464732-5G
77484-010EA 342.99 USD
77484-010 77494-480
1-(2-Fluoro-5-methoxyphenyl)propan-1-one ≥95%
1-(2-Fluoro-5-methoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
CAS Number: 89106-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR