4-Chloro-2-iodobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

464700-500MG 464700-5G 464700-25G
77484-006EA 99.91 USD
77484-006 77494-476 77504-098
4-Chloro-2-iodobenzo[b]thiophene ≥95%
4-Chloro-2-iodobenzo[b]thiophene

Precaución: For research use only

Formula: C₈H₄ClIS
MW: 294.54 g/mol
Storage Temperature: Ambient
CAS Number: 3031788-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR