(4-Bromo-5-methyl-2-thienyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

464358-500MG 464358-5G
77484-002EA 408.91 USD
77484-002 77494-472
(4-Bromo-5-methyl-2-thienyl)cyclopropylmethanone ≥95%
(4-Bromo-5-methyl-2-thienyl)cyclopropylmethanone

Precaución: For research use only

Formula: C₉H₉BrOS
MW: 245.14 g/mol
Storage Temperature: Ambient
CAS Number: 1595712-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR