4-Bromo-α-ethenyl-5-methyl-2-thiophenemethanol ≥95%

Supplier: AOBChem USA

Warning

464332-500MG 464332-5G
77483-998EA 505.73 USD
77483-998 77494-468
4-Bromo-α-ethenyl-5-methyl-2-thiophenemethanol ≥95%
4-Bromo-α-ethenyl-5-methyl-2-thiophenemethanol

Precaución: For research use only

Formula: C₈H₉BrOS
Storage Temperature: Ambient
CAS Number: 1593015-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR