6-Isopropoxypicolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

46361-500MG 46361-5G 46361-25G
77483-994EA 184.37 USD
77483-994 77494-464 77504-094
6-Isopropoxypicolinaldehyde ≥97%
6-Isopropoxypicolinaldehyde

Precaución: For research use only

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD11977419
CAS Number: 350697-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR