5-Bromo-2,3-difluoro-4-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

46313-500MG 46313-5G
77483-990EA 416.12 USD
77483-990 77494-460
5-Bromo-2,3-difluoro-4-methoxybenzonitrile ≥97%
5-Bromo-2,3-difluoro-4-methoxybenzonitrile

Precaución: For research use only

Formula: C₈H₄BrF₂NO
MW: 248.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34813529
CAS Number: 1897939-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR