(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

46135-500MG 46135-5G 46135-25G
77483-986EA 119.48 USD
77483-986 77494-456 77504-090
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-2-bromo-6-fluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₂BrFOS
Storage Temperature: Ambient
MDL Number: MFCD32707157
CAS Number: 2432848-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR