2-Methoxy-1-(o-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

460679-500MG 460679-5G 460679-25G
77483-982EA 305.91 USD
77483-982 77494-452 77504-086
2-Methoxy-1-(o-tolyl)ethanone ≥95%
2-Methoxy-1-(o-tolyl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₂O₂
MW: 164.2010 g/mol
Storage Temperature: Ambient
CAS Number: 97728-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR