tert-Butyl 6-phenylnicotinate ≥95%

Supplier: AOBChem USA

Warning

460635-500MG 460635-5G 460635-25G
77483-978EA 141.11 USD
77483-978 77494-448 77504-082
tert-Butyl 6-phenylnicotinate ≥95%
tert-Butyl 6-phenylnicotinate

Precaución: For research use only

Formula: C₁₆H₁₇NO₂
Storage Temperature: Ambient
CAS Number: 89808-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR