1-(4-(cyclohexyloxy)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

458776-500MG 458776-5G 458776-25G
77483-968EA 296.64 USD
77483-968 77494-436 77504-074
1-(4-(cyclohexyloxy)phenyl)propan-1-one ≥95%
1-(4-(cyclohexyloxy)phenyl)propan-1-one

Precaución: For research use only

Formula: C₁₅H₂₀O₂
MW: 232.318 g/mol
Storage Temperature: Ambient
CAS Number: 101498-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR