(2-Fluoro-3-(trifluoromethoxy)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

457919-500MG 457919-5G 457919-25G
77483-964EA 296.64 USD
77483-964 77494-432 77504-070
(2-Fluoro-3-(trifluoromethoxy)phenyl)(phenyl)methanone ≥95%
(2-Fluoro-3-(trifluoromethoxy)phenyl)(phenyl)methanone

Precaución: For research use only

Formula: C₁₄H₈F₄O₂
Storage Temperature: Ambient
CAS Number: 3031808-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR