(3-Bromo-2-fluoro-5-methylphenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

457602-500MG 457602-5G
77483-956EA 390.37 USD
77483-956 77494-424
(3-Bromo-2-fluoro-5-methylphenyl)(cyclopropyl)methanone ≥95%
(3-Bromo-2-fluoro-5-methylphenyl)(cyclopropyl)methanone

Precaución: For research use only

Formula: C₁₁H₁₀BrFO
MW: 257.099 g/mol
Storage Temperature: Ambient
CAS Number: 2293239-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR