benzofuran-2-yl(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

457131-500MG 457131-5G 457131-25G
77483-952EA 202.91 USD
77483-952 77494-420 77504-060
benzofuran-2-yl(cyclopropyl)methanol ≥95%
benzofuran-2-yl(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₂O₂
Storage Temperature: Ambient
CAS Number: 1339173-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR