Ethyl 2-(2,4-dichlorothiazol-5-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

456885-500MG 456885-5G
77483-948EA 528.39 USD
77483-948 77494-416
Ethyl 2-(2,4-dichlorothiazol-5-yl)-2-oxoacetate ≥95%
Ethyl 2-(2,4-dichlorothiazol-5-yl)-2-oxoacetate

Precaución: For research use only

Formula: C₇H₅Cl₂NO₃S
MW: 254.0905 g/mol
Storage Temperature: Ambient
CAS Number: 2733594-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR