2-(4-(tert-Butyl)phenyl)-6-methylpyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
45598-500MG
45598-5G
45598-25G
77483-946EA
202.91
USD
77483-946
77494-412
77504-056
2-(4-(tert-Butyl)phenyl)-6-methylpyridine ≥95%
2-(4-(tert-Butyl)phenyl)-6-methylpyridine
Precaución: For research use only
Formula:
C₁₆H₁₉N MW: 225.329 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28340573 CAS Number: 1226769-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...