2-(2-Bromo-6-chloropyridin-4-yl)acetic acid ≥95%

Supplier: AOBChem USA

Warning

452768-500MG 452768-5G
77483-936EA 367.71 USD
77483-936 77494-400
2-(2-Bromo-6-chloropyridin-4-yl)acetic acid ≥95%
2-(2-Bromo-6-chloropyridin-4-yl)acetic acid

Precaución: For research use only

Formula: C₇H₅BrClNO₂
MW: 250.4771 g/mol
Storage Temperature: Ambient
CAS Number: 1227593-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR