2-(Benzofuran-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

452154-1G 452154-500MG 452154-5G 452154-25G
77471-280EA 258.53 USD
77471-280 77483-932 77494-396 77504-044
2-(Benzofuran-2-yl)propan-2-ol ≥95%
2-(Benzofuran-2-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₂O₂
MW: 176.21 g/mol
Storage Temperature: Ambient
CAS Number: 59302-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR