1-(2-Methylnaphthalen-1-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

449009-500MG 449009-5G 449009-25G
77483-928EA 258.53 USD
77483-928 77494-392 77504-040
1-(2-Methylnaphthalen-1-yl)cyclobutanol ≥95%
1-(2-Methylnaphthalen-1-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₅H₁₆O
Storage Temperature: Ambient
CAS Number: 2747206-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR