tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl 5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylate

44340-500MG 44340-5G 44340-25G
77483-904EA 130.81 USD
77483-904 77494-370 77504-024
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate ≥95%
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

Precaución: For research use only

Formula: C₁₆H₂₄BNO₄
MW: 305.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26399628
CAS Number: 1354356-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR