3-Pyridinecarboxylic acid, 2-bromo-5-methyl- ≥95%

Supplier: AOBChem USA

Warning

439911-500MG 439911-5G 439911-25G
77483-896EA 182.31 USD
77483-896 77494-362 77504-016
3-Pyridinecarboxylic acid, 2-bromo-5-methyl- ≥95%
3-Pyridinecarboxylic acid, 2-bromo-5-methyl-

Precaución: For research use only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Storage Temperature: Ambient
CAS Number: 65996-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR