3-Methoxy-n,n-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%
Supplier: AOBChem USA
|
Warning
|
43128-500MG
43128-5G
77483-880EA
381.1
USD
77483-880
77494-346
3-Methoxy-n,n-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%
3-Methoxy-n,n-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Precaución: For research use only
Formula:
C₁₆H₂₄BNO₄ MW: 305.177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18760158 CAS Number: 2059087-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...