1-[2-(1-Methylethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

430984-500MG 430984-5G
77483-876EA 463.5 USD
77483-876 77494-342
1-[2-(1-Methylethyl)phenyl]-1-propanone ≥95%
1-[2-(1-Methylethyl)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
CAS Number: 107075-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR