Ethyl 2-(5-(tert-butyl)-2-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

430108-500MG 430108-5G 430108-25G
77483-872EA 296.64 USD
77483-872 77494-338 77504-006
Ethyl 2-(5-(tert-butyl)-2-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-(tert-butyl)-2-methoxyphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₅H₂₀O₄
Storage Temperature: Ambient
CAS Number: 1225542-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR